siRNA Details
Gene Symbol: PAGE2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015038.2 | NP_001015038.1 | 5 | 444 | ||
XM_011530785.2 | XP_011529087.1 | 6 | 860 | |||
XM_011530786.2 | XP_011529088.1 | 5 | 527 | |||
XM_011530787.2 | XP_011529089.1 | 5 | 507 | |||
XM_017029513.1 | XP_016885002.1 | 5 | 464 | |||
GenBank mRNA | BM782405.1 | - | 5 | 429 |
Gene Symbol: PAGE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207339.3 | NP_997222.1 | 5 | 437 | ||
XM_017029353.1 | XP_016884842.1 | 5 | 466 | |||
GenBank mRNA | BC054022.1 | - | 5 | 431 | 6603202 | |
BX113994.1 | - | 5 | 367 |
