siRNA Details
Gene Symbol: KANK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198471.2 | NP_940873.2 | 11 | 2549 | ||
XM_006722718.3 | XP_006722781.1 | 11 | 2668 | |||
XM_011527884.2 | XP_011526186.1 | 11 | 2925 | |||
XM_017026563.1 | XP_016882052.1 | 10 | 2685 | |||
GenBank mRNA | AB284126.1 | - | 11 | 2549 | ||
AK128815.1 | - | 11 | 2549 | |||
BC066124.1 | - | 11 | 2549 | 30345663 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
