siRNA Details
Gene Symbol: SVIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320340.1 | NP_001307269.1 | 3 | 351 | ||
NM_001320341.1 | NP_001307270.1 | 3 | 242 | |||
NM_001320342.1 | NP_001307271.1 | 3 | 242 | |||
NM_148893.2 | NP_683691.1 | 3 | 242 | |||
NR_135213.1 | 3 | 197 | ||||
GenBank mRNA | AA347778.1 | - | 3 | 193 | ||
AF527534.1 | - | 3 | 216 | |||
AL833119.1 | - | 3 | 215 | |||
BF219509.1 | - | 3 | 199 | |||
BG531443.1 | - | 3 | 175 | |||
BI826588.1 | - | 3 | 350 | |||
DA306370.1 | - | 3 | 192 | |||
DB135755.1 | - | 3 | 245 | |||
DB466086.1 | - | 3 | 178 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
