siRNA Details
Gene Symbol: ZNF843 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136509.1 | NP_001129981.1 | 2 | 1960 | ||
GenBank mRNA | AK291063.1 | - | 2 | 1718 | ||
BC036762.1 | - | 2 | 1586 | 5588928 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
