siRNA Details
Gene Symbol: LOC284009 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028335.1 | 3 | 519 | |||
GenBank mRNA | AK093990.1 | - | 3 | 1166 | ||
BC093711.1 | - | 3 | 520 | 7939556 | ||
BC112034.1 | - | 3 | 520 | 8327502 |
