siRNA Details
Gene Symbol: SPDYE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175064.2 | NP_778234.2 | 5 | 958 | ||
XM_005249719.3 | XP_005249776.1 | 7 | 1469 | |||
XM_017012015.1 | XP_016867504.1 | 7 | 1407 | |||
XM_017012016.1 | XP_016867505.1 | 6 | 819 | |||
XM_017012017.1 | XP_016867506.1 | 4 | 1054 | |||
GenBank mRNA | AF412027.1 | - | 5 | 967 | ||
AL137266.1 | - | 5 | 740 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
