siRNA Details
Gene Symbol: CLPSL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010886.3 | NP_001010886.1 | 3 | 374 | ||
XM_017010820.1 | XP_016866309.1 | 2 | 453 | |||
GenBank mRNA | BC133046.1 | - | 3 | 374 | 40147068 | |
BC137392.1 | - | 3 | 374 | 9021390 | ||
BC150543.1 | - | 3 | 376 | 40127356 | ||
BC150544.1 | - | 3 | 376 | 40127359 | ||
BX106742.1 | - | 3 | 384 |
