siRNA Details
Gene Symbol: PFN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029886.2 | NP_001025057.1 | 1 | 425 | ||
GenBank mRNA | BC132952.1 | - | 1 | 425 | 40146974 | |
BC132954.1 | - | 1 | 425 | 40146976 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
