siRNA Details
Gene Symbol: KRTAP10-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181688.2 | NP_859016.1 | 1 | 517 | ||
GenBank mRNA | AB076358.1 | - | 1 | 502 | ||
AJ566387.1 | - | 1 | 517 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: KRTAP10-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181688.2 | NP_859016.1 | 1 | 517 | ||
GenBank mRNA | AB076358.1 | - | 1 | 502 | ||
AJ566387.1 | - | 1 | 517 |