siRNA Details
Gene Symbol: KRTAP10-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198687.2 | NP_941960.2 | 1 | 1338 | ||
GenBank mRNA | AB076351.1 | - | 1 | 1338 | ||
AJ566382.1 | - | 1 | 99 | |||
BC125048.1 | - | 1 | 1327 | 40116133 | ||
BC125049.1 | - | 1 | 1327 | 40116140 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
