siRNA Details
Gene Symbol: KRTAP10-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198689.2 | NP_941962.1 | 1 | 210 | ||
GenBank mRNA | AB076354.1 | - | 1 | 210 | ||
BC119643.2 | - | 1 | 207 | 40115724 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
