siRNA Details
Gene Symbol: KRTAP10-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB076356.1 | - | 1 | 1020 | ||
AJ566386.1 | - | 1 | 244 | |||
BC131613.1 | - | 1 | 996 | 40115414 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
