siRNA Details
Gene Symbol: KRTAP10-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198692.2 | NP_941965.2 | 1 | 946 | ||
GenBank mRNA | AB076359.1 | - | 1 | 946 | ||
AF086314.1 | - | 1 | 126 | |||
BC131611.1 | - | 1 | 927 | 40115290 | ||
BX103563.1 | - | 1 | 124 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
