siRNA Details
Gene Symbol: TMEM233 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005253881.3 | XP_005253938.1 | 3 | 2058 | ||
XM_011538328.2 | XP_011536630.1 | 3 | 2059 | |||
XM_011538329.2 | XP_011536631.1 | 3 | 2059 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
