siRNA Details
Gene Symbol: C2CD4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007595.2 | NP_001007596.2 | 2 | 265 | ||
XM_017022170.1 | XP_016877659.1 | 3 | 606 | |||
GenBank mRNA | BM023530.1 | - | 1 | 162 | ||
BM127548.1 | - | 2 | 222 | |||
BM850957.1 | - | 2 | 267 | |||
BQ101161.1 | - | 2 | 252 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
