siRNA Details
Gene Symbol: LOC388780 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287682.1 | NP_001274611.1 | 2 | 606 | ||
XM_017027847.1 | XP_016883336.1 | 2 | 1566 | |||
XM_017027848.1 | XP_016883337.1 | 2 | 1584 |
