siRNA Details
Gene Symbol: C2orf82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_206895.1 | NP_996778.1 | 1, 2 | 134 | ||
XM_017004080.1 | XP_016859569.1 | 2, 3 | 1164 | |||
XM_017004081.1 | XP_016859570.1 | 2, 3 | 902 | |||
XM_017004082.1 | XP_016859571.1 | 3, 4 | 1222 | |||
XM_017004083.1 | XP_016859572.1 | 2, 3 | 934 | |||
XM_017004084.1 | XP_016859573.1 | 2, 3 | 197 | |||
GenBank mRNA | AY358535.1 | - | 1, 2 | 134 | ||
BC130307.1 | - | 1, 2 | 132 | 40146338 | ||
BC130309.1 | - | 1, 2 | 132 | 40146340 |
