siRNA Details
Gene Symbol: OR6B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005853.1 | NP_001005853.1 | 1 | 853 | ||
GenBank mRNA | BC137561.1 | - | 1 | 913 | 9021561 | |
BC137562.1 | - | 1 | 913 | 9021562 |
