siRNA Details
Gene Symbol: LRRC26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013653.2 | NP_001013675.1 | 2 | 844 | ||
GenBank mRNA | BC140911.1 | - | 2 | 813 | 9056723 | |
CB160046.1 | - | 2 | 224 | |||
DQ355157.1 | - | 2 | 780 | |||
EU588721.1 | - | 2 | 235 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
