siRNA Details
Gene Symbol: LOC390877 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321601.1 | NP_001308530.1 | 1 | 67 | ||
XM_017026804.1 | XP_016882293.1 | 1 | 194 | |||
XM_017026805.1 | XP_016882294.1 | 2 | 661 | |||
GenBank mRNA | BC157867.1 | - | 1 | 81 | ||
BC171868.1 | - | 1 | 81 |
