siRNA Details
Gene Symbol: TBC1D3F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK292344.1 | - | 14 | 2061 | ||
BC075809.1 | - | 14 | 2050 | 5170064 |
Gene Symbol: TBC1D3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001417.6 | NP_001001417.6 | 14 | 2050 | ||
GenBank mRNA | AF540953.1 | - | 13 | 1904 | ||
AK292257.1 | - | 14 | 2021 |
Gene Symbol: TBC1D3P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027486.1 | 14 | 2005 | |||
GenBank mRNA | BC058890.1 | - | 14 | 2030 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
