siRNA Details
Gene Symbol: FAM183A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005270875.4 | XP_005270932.1 | 3 | 1185 | ||
XM_005270876.4 | XP_005270933.1 | 3 | 2533 | |||
XM_017001322.1 | XP_016856811.1 | 4 | 2411 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
