siRNA Details
Gene Symbol: SPDYE16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006716211.3 | XP_006716274.1 | 5 | 1090 | ||
XM_017012887.1 | XP_016868376.1 | 5 | 1090 |
Gene Symbol: SPDYE18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005250742.4 | XP_005250799.1 | 6 | 961 | ||
XM_005250743.4 | XP_005250800.1 | 5 | 816 | |||
XM_011516756.2 | XP_011515058.1 | 5 | 730 |
Gene Symbol: SPDYE7P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003666.2 | 4 | 648 | |||
GenBank mRNA | BC100974.2 | - | 4 | 648 |
