siRNA Details
Gene Symbol: TRIM6-TRIM34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003819.3 | NP_001003819.1 | 7, 8 | 1244 | ||
GenBank mRNA | AB039903.1 | - | 7, 8 | 1152 | ||
BC136871.1 | - | 7, 8 | 1163 | 9020862 | ||
BC140722.1 | - | 7, 8 | 1163 | 9021590 | ||
BC144424.1 | - | 7, 8 | 1163 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
