siRNA Details
Gene Symbol: C1QTNF9B-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014442.2 | NP_001014442.2 | 4 | 761 | ||
NM_001135816.1 | NP_001129288.1 | 4 | 658 | |||
GenBank mRNA | BC015452.1 | - | 4 | 763 | ||
BC073902.1 | - | 4 | 761 | |||
BC142999.1 | - | 4 | 658 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
