siRNA Details
Gene Symbol: OVOL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302757.1 | NP_001289686.1 | 2 | 129 | ||
XM_011527251.2 | XP_011525553.1 | 1 | 23 | |||
XM_017027190.1 | XP_016882679.1 | 1 | 224 | |||
XM_017027191.1 | XP_016882680.1 | 2 | 152 |
