siRNA Details
Gene Symbol: SMIM10L2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024359.2 | 1 | 386 | |||
GenBank mRNA | BC030620.1 | - | 1 | 385 | ||
BC051704.1 | - | 1 | 373 |
Gene Symbol: SMIM10L2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024493.2 | 1 | 188 | |||
GenBank mRNA | AK123302.1 | - | 1 | 188 |
