siRNA Details
Gene Symbol: RNF225 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195135.1 | NP_001182064.1 | 1 | 263 | ||
XM_006723328.3 | XP_006723391.1 | 2 | 894 |

More Information
Other Species Targeted

Gene Ontology Categories:
