siRNA Details
Gene Symbol: LOC81787 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL512723.1 | - | 1 | 1572 |
Gene Symbol: LOC107987377 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017030113.1 | XP_016885602.1 | 4 | 2176 |
