siRNA Details
Gene Symbol: PRB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006248.3 | NP_006239.3 | 3 | 550 | ||
GenBank mRNA | K03208.1 | - | 1 | 20 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
