siRNA Details
Gene Symbol: SPAG11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081552.2 | NP_001075021.2 | 2, 3 | 392 | ||
XM_005272391.4 | XP_005272448.2 | 2, 3 | 365 | |||
XM_005272393.4 | XP_005272450.2 | 3, 4 | 441 | |||
XM_017013744.1 | XP_016869233.1 | 1, 2 | 66 | |||
XM_017013745.1 | XP_016869234.1 | 2, 3 | 142 | |||
GenBank mRNA | AF170797.1 | - | 2, 3 | 204 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
