siRNA Details
Gene Symbol: SCGB1C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145651.2 | NP_663626.2 | 3 | 308 | ||
XM_005252804.3 | XP_005252861.2 | 4 | 588 | |||
GenBank mRNA | AY026938.1 | - | 3 | 308 | ||
BC069287.1 | - | 3 | 308 | 7262728 | ||
BC111427.1 | - | 3 | 307 | 40069184 | ||
BC111433.1 | - | 3 | 307 | 40069182 |
Gene Symbol: SCGB1C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001097610.2 | NP_001091079.1 | 3 | 308 | ||
XM_011535362.2 | XP_011533664.1 | 4 | 588 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
