siRNA Details
Gene Symbol: FAM72B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100910.1 | NP_001094380.1 | 4 | 1680 | ||
NM_001320149.1 | NP_001307078.1 | 4 | 1153 | |||
XM_011542018.1 | XP_011540320.1 | 5 | 2088 | |||
GenBank mRNA | BC046199.1 | - | 4 | 1153 | 6064384 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
