siRNA Details
Gene Symbol: FRMPD2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033172.1 | 2 | 464 | |||
GenBank mRNA | AK294650.1 | - | 2 | 419 | ||
AK313659.1 | - | 2 | 421 | |||
AK315971.1 | - | 2 | 464 | |||
BC071635.1 | - | 2 | 376 | |||
BC072395.1 | - | 2 | 390 | |||
BC110798.1 | - | 2 | 387 | |||
BC137431.1 | - | 2 | 351 | 9021429 | ||
BC137434.1 | - | 2 | 351 | 9021432 | ||
BC137439.1 | - | 2 | 349 | 9021437 | ||
BC137440.1 | - | 2 | 351 | 9021438 |
Gene Symbol: FRMPD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK123038.1 | - | 23 | 3183 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
