siRNA Details
Gene Symbol: LOC101929950 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006722258.3 | XP_006722321.1 | 5 | 565 | ||
XM_017025500.1 | XP_016880989.1 | 5 | 570 |
Gene Symbol: LOC440434 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036750.2 | 6 | 763 |
