siRNA Details
Gene Symbol: LOC100996318 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003846491.4 | XP_003846539.1 | 3 | 892 |
Gene Symbol: HIST2H2BF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK092418.1 | - | 1 | 1270 |
Gene Symbol: LOC100996713 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006711719.3 | XP_006711782.1 | 2 | 817 |
