siRNA Details
Gene Symbol: LINC00965 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027000.1 | 1 | 1692 | |||
GenBank mRNA | AK090418.1 | - | 1 | 3679 | ||
BC093747.1 | - | 1 | 275 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00965 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027000.1 | 1 | 1692 | |||
GenBank mRNA | AK090418.1 | - | 1 | 3679 | ||
BC093747.1 | - | 1 | 275 |