siRNA Details
Gene Symbol: LINC01136 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034150.1 | 2, 3 | 661 | |||
NR_034151.1 | 3, 4 | 847 | ||||
GenBank mRNA | AK024293.1 | - | 2, 3 | 645 | ||
AK309664.1 | - | 3, 4 | 831 |
