siRNA Details
Gene Symbol: ZNF730 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277403.1 | NP_001264332.1 | 4 | 849 | ||
XM_017026114.1 | XP_016881603.1 | 4 | 766 | |||
XM_017026115.1 | XP_016881604.1 | 4 | 926 | |||
GenBank mRNA | AK131472.1 | - | 4 | 849 |
