siRNA Details
Gene Symbol: SPDYE18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005250742.4 | XP_005250799.1 | 1 | 36 | ||
XM_011516756.2 | XP_011515058.1 | 1 | 36 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SPDYE18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005250742.4 | XP_005250799.1 | 1 | 36 | ||
XM_011516756.2 | XP_011515058.1 | 1 | 36 |