siRNA Details
Gene Symbol: GTF2H2C_2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042490.3 | NP_001035955.1 | 9 | 739 | ||
GenBank mRNA | BC064557.1 | - | 8 | 619 | 5549265 | |
BC065021.1 | - | 9 | 740 | 6047020 | ||
BX647445.1 | - | 8 | 504 |
Gene Symbol: GTF2H2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033417.1 | 7 | 628 | |||
GenBank mRNA | BC171919.1 | - | 7 | 557 | 9054573 |
Gene Symbol: GTF2H2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098728.1 | NP_001092198.1 | 8 | 664 | ||
XM_005248588.4 | XP_005248645.1 | 8 | 591 | |||
XM_005248590.3 | XP_005248647.1 | 8 | 529 | |||
XM_005248592.3 | XP_005248649.1 | 6 | 393 | |||
XM_006714688.3 | XP_006714751.1 | 8 | 582 | |||
XM_011543615.1 | XP_011541917.1 | 8 | 664 | |||
XM_017009804.1 | XP_016865293.1 | 6 | 447 | |||
GenBank mRNA | AK292361.1 | - | 8 | 643 | ||
AK315282.1 | - | 8 | 664 | |||
BC105980.1 | - | 8 | 522 | 4445643 | ||
BC157894.1 | - | 8 | 594 | |||
BC171860.1 | - | 9 | 656 | 9054514 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
