siRNA Details
Gene Symbol: KLK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030047.1 | NP_001025218.1 | 2, 3 | 253 | ||
NM_001648.2 | NP_001639.1 | 2, 3 | 253 | |||
GenBank mRNA | AF335477.1 | - | 2, 3 | 676 | ||
AF335478.1 | - | 1, 2 | 1491 | |||
AJ310937.1 | - | 2, 3 | 249 | |||
AJ310938.1 | - | 2, 3 | 249 | |||
AJ459782.1 | - | 2, 3 | 233 | |||
AJ459783.1 | - | 2, 3 | 233 | |||
AJ512346.1 | - | 2, 3 | 233 | |||
AK312933.1 | - | 2, 3 | 247 | |||
AX772834.1 | - | 2, 4 | 253 | |||
BC005307.1 | - | 2, 3 | 253 | 3950475 | ||
BC050595.2 | - | 2, 3 | 276 | 6199928 | ||
BC056665.1 | - | 2, 3 | 258 | 4296876 | ||
BT019862.1 | - | 2, 3 | 212 | |||
M21895.1 | - | 2, 3 | 201 | |||
M21896.1 | - | 2, 3 | 201 | |||
M21897.1 | - | 2, 3 | 201 | |||
M26663.1 | - | 2, 3 | 236 | |||
S75755.1 | - | 2, 3 | 253 | |||
U17040.1 | - | 2, 3 | 253 | |||
X05332.1 | - | 2, 3 | 255 | |||
X07730.1 | - | 1, 2 | 517 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
