siRNA Details
Gene Symbol: ABCC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171.5 | NP_001162.4 | 19 | 2510 | ||
XM_011522479.2 | XP_011520781.1 | 19 | 2621 | |||
XM_011522480.1 | XP_011520782.1 | 19 | 2458 | |||
XM_011522481.2 | XP_011520783.1 | 19 | 2657 | |||
XM_011522482.2 | XP_011520784.1 | 19 | 2651 | |||
XM_017023212.1 | XP_016878701.1 | 18 | 2486 | |||
XM_017023213.1 | XP_016878702.1 | 19 | 2544 | |||
XM_017023214.1 | XP_016878703.1 | 19 | 2655 | |||
GenBank mRNA | AF076622.1 | - | 19 | 2496 | ||
AF168791.1 | - | 19 | 2473 | |||
AM774324.1 | - | 19 | 2473 | |||
BC131732.1 | - | 19 | 2496 | 40124570 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
