siRNA Details
Gene Symbol: CHRM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000740.2 | NP_000731.1 | 5 | 1149 | ||
XM_005273032.3 | XP_005273089.1 | 6 | 1454 | |||
XM_011544041.2 | XP_011542343.1 | 7 | 1553 | |||
XM_011544043.2 | XP_011542345.1 | 9 | 1886 | |||
XM_011544044.2 | XP_011542346.1 | 8 | 1657 | |||
XM_011544045.2 | XP_011542347.1 | 8 | 1782 | |||
XM_011544046.2 | XP_011542348.1 | 6 | 1490 | |||
XM_011544047.2 | XP_011542349.1 | 6 | 1426 | |||
XM_017000152.1 | XP_016855641.1 | 8 | 4479 | |||
XM_017000153.1 | XP_016855642.1 | 7 | 1557 | |||
XM_017000154.1 | XP_016855643.1 | 6 | 1251 | |||
XM_017000155.1 | XP_016855644.1 | 7 | 1594 | |||
XM_017000156.1 | XP_016855645.1 | 7 | 1558 | |||
XM_017000157.1 | XP_016855646.1 | 5 | 1159 | |||
XM_017000158.1 | XP_016855647.1 | 5 | 1391 | |||
XM_017000159.1 | XP_016855648.1 | 5 | 1363 | |||
XM_017000160.1 | XP_016855649.1 | 4 | 2213 | |||
XM_017000161.1 | XP_016855650.1 | 6 | 1495 | |||
XM_017000162.1 | XP_016855651.1 | 5 | 1327 | |||
XM_017000163.1 | XP_016855652.1 | 4 | 5189 | |||
GenBank mRNA | AF279779.1 | - | 5 | 1148 | ||
AF385589.1 | - | 1 | 370 | |||
AF498917.1 | - | 1 | 370 | |||
AK074575.1 | - | 1 | 527 | |||
BC096844.1 | - | 1 | 371 | |||
BC121026.2 | - | 2 | 513 | 40117429 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
