siRNA Details
Gene Symbol: ABCC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105515.2 | NP_001098985.1 | 10 | 1428 | ||
NM_001301829.1 | NP_001288758.1 | 10 | 1428 | |||
NM_001301830.1 | NP_001288759.1 | 9 | 1203 | |||
NM_005845.4 | NP_005836.2 | 10 | 1428 | |||
XM_005254025.2 | XP_005254082.1 | 11 | 1488 | |||
XM_017020319.1 | XP_016875808.1 | 11 | 1574 | |||
XM_017020320.1 | XP_016875809.1 | 10 | 1532 | |||
XM_017020322.1 | XP_016875811.1 | 11 | 1437 | |||
GenBank mRNA | AB208973.1 | - | 8 | 1047 | ||
AF071202.1 | - | 10 | 1411 | |||
AF071203.1 | - | 2 | 66 | |||
AF541977.1 | - | 10 | 1301 | |||
AK290317.1 | - | 10 | 1428 | |||
AK296247.1 | - | 9 | 1190 | |||
AK298953.1 | - | 10 | 1392 | |||
AY081219.1 | - | 10 | 1317 | |||
AY133678.1 | - | 11 | 1470 | |||
AY133679.1 | - | 12 | 1583 | |||
AY133680.1 | - | 11 | 1430 | |||
AY207008.1 | - | 10 | 1301 | |||
BC041560.1 | - | 10 | 1398 | 5812052 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
