siRNA Details
Gene Symbol: DNMT3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320893.1 | NP_001307822.1 | 16 | 2010 | ||
NM_022552.4 | NP_072046.2 | 21 | 2696 | |||
NM_153759.3 | NP_715640.2 | 17 | 1980 | |||
NM_175629.2 | NP_783328.1 | 21 | 2767 | |||
NR_135490.1 | 22 | 2966 | ||||
XM_005264175.4 | XP_005264232.1 | 21 | 2630 | |||
XM_005264177.4 | XP_005264234.1 | 16 | 1922 | |||
XM_011532662.2 | XP_011530964.1 | 19 | 2632 | |||
XM_011532663.2 | XP_011530965.1 | 22 | 2809 | |||
XM_011532666.2 | XP_011530968.1 | 17 | 1995 | |||
XM_011532667.2 | XP_011530969.1 | 16 | 1812 | |||
XM_017003526.1 | XP_016859015.1 | 21 | 2738 | |||
XM_017003527.1 | XP_016859016.1 | 16 | 1966 | |||
GenBank mRNA | AB208833.1 | - | 22 | 2865 | ||
AF067972.2 | - | 21 | 2658 | |||
AF331856.1 | - | 21 | 2646 | |||
AF480163.1 | - | 17 | 1976 | |||
AK025230.1 | - | 11 | 1020 | |||
BC018214.1 | - | 6 | 549 | |||
BC023612.2 | - | 7 | 748 | |||
BC043617.1 | - | 21 | 2666 | 6150112 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
