siRNA Details
Gene Symbol: EDA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005609.1 | NP_001005609.1 | 7, 8 | 1153 | ||
NM_001005612.2 | NP_001005612.2 | 7, 8 | 1144 | |||
NM_001399.4 | NP_001390.1 | 7 | 1153 | |||
XM_006724630.2 | XP_006724693.1 | 7 | 1125 | |||
GenBank mRNA | AF040628.1 | - | 7 | 1153 | ||
AF060999.1 | - | 7 | 1152 | |||
AF061189.1 | - | 7, 8 | 1153 | |||
BC126143.1 | - | 7 | 1103 | 8991859 | ||
BC144049.1 | - | 7, 8 | 1094 | 9052569 | ||
BC144051.1 | - | 7, 8 | 1103 | 9052571 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
