siRNA Details
Gene Symbol: GH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000515.4 | NP_000506.2 | 3 | 347 | ||
NM_022559.3 | NP_072053.1 | 3 | 302 | |||
GenBank mRNA | A00469.1 | - | 3 | 310 | ||
AF110644.1 | - | 3 | 281 | |||
AF185611.1 | - | 3 | 289 | |||
AV749353.1 | - | 3 | 347 | |||
AV753220.1 | - | 1 | 167 | |||
AY613431.1 | - | 3 | 333 | |||
AY613432.1 | - | 3 | 333 | |||
BC062475.1 | - | 3 | 267 | 30393581 | ||
BC075012.2 | - | 3 | 298 | 30915404 | ||
BC075013.2 | - | 3 | 298 | 30915617 | ||
BC090045.1 | - | 3 | 314 | 30394115 | ||
V00519.1 | - | 3 | 311 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
