siRNA Details
Gene Symbol: GH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022560.3 | NP_072054.1 | 2, 3 | 250 | ||
GenBank mRNA | CD108592.1 | - | 2, 3 | 216 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
