siRNA Details
Gene Symbol: HFE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF115264.1 | - | 4 | 443 | ||
AF144240.1 | - | 3 | 645 | |||
AJ404378.1 | - | 2 | 321 | |||
U60319.1 | - | 7 | 2436 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
